Search
Search
| Gene Symbol: Toe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026654.2 | NP_080930.1 | 3 - 4 | 278 | 81 | |
| GenBank mRNA | AK016889.1 | - | 268 | 81 | ||
| AK019829.1 | - | 3 - 4 | 278 | 81 | ||
| AK046576.1 | - | 3 - 4 | 309 | 81 | ||
| AK082975.1 | - | 3 - 4 | 259 | 81 | ||
| BC023109.1 | - | 3 - 4 | 254 | 4021435 | 81 | |
Set Membership: |
|