Search
Search
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013484.2 | NP_038512.2 | 6 - 7 | 974 | 101 | |
| GenBank mRNA | AK146812.1 | - | 6 - 7 | 975 | 101 | |
| BC011086.1 | - | 6 - 7 | 937 | 3992881 | 101 | |
Set Membership: |
|