Search
Search

Arid4b| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194262.2 | NP_919238.1 | 19 - 20 | 2424 | 125 | |
| NM_198122.2 | NP_937755.1 | 18 - 19 | 2163 | 125 | ||
| GenBank mRNA | AK018194.1 | - | 2 - 3 | 3527 | 125 | |
| AK080967.1 | - | 3 - 4 | 325 | 125 | ||
| AK163022.1 | - | 2 - 3 | 913 | 125 | ||
| BC024724.1 | - | 4 - 5 | 702 | 125 | ||
| BC137783.1 | - | 18 - 19 | 2186 | 8860802 | 125 | |
Set Membership: |
|