Search
Search

Abcb8| Gene Symbol: Abcb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029020.2 | NP_083296.2 | 9 - 10 | 1558 | 68 | |
| GenBank mRNA | AK014319.1 | - | 9 - 10 | 1289 | 68 | |
| AK030624.1 | - | 9 - 10 | 1285 | 68 | ||
| AK038760.1 | - | 9 - 10 | 1286 | 68 | ||
| AK049152.1 | - | 9 - 10 | 1302 | 68 | ||
| AK076315.1 | - | 9 - 10 | 1309 | 68 | ||
| AK166704.1 | - | 9 - 10 | 1292 | 68 | ||
| BC015301.1 | - | 9 - 10 | 1558 | 4237894 | 68 | |
Set Membership: |
|