Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01135138_g1
          See other SPACA6 GE Assays ›
          Gene Symbol:
          Spaca6
          Gene Name:
          sperm acrosome associated 6
          Gene Aliases:
          4930546H06Rik, 9530079D04Rik, B230206P06Rik, Ncrna00085, let-7e, mir-125a, mir-99b
          Chromosome Location:
          Chr.17: 17824821 - 17843010 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01135138_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spaca6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162909.1 NP_001156381.1 8 - 9 1016 93
          GenBank mRNA AK080293.1 - 3 - 4 453 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000211.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          fusion of sperm to egg plasma membrane
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017570 AK080293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017570 BC171977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017570 BC171982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017570 BC171984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017570 NM_001162909.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173329 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017570 AK080293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017570 BC171977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017570 BC171982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017570 BC171984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017570 NM_001162909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357962 BC171977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357962 BC171982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357962 NM_001162909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P357962 AK016052.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442090 - -
          Affymetrix HT MG-430 PM Array Plate 1430959_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453713_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1458116_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442087 - -
          Mouse Genome 430 2.0 Array 1442335_at - -
          Mouse Genome 430 2.0 Array 1430959_at - -
          Mouse Genome 430 2.0 Array 1430959_PM_at - -
          Mouse Genome 430 2.0 Array 1453713_s_at - -
          Mouse Genome 430 2.0 Array 1453713_PM_s_at - -
          Mouse Genome 430 2.0 Array 1442335_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75202_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75202_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700001711.mm.1 NM_001162909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001711.mm.1 NM_001162909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001711.mm.1 TC1700000211.mm.1 MTA TC
          Mouse Expression Set 430 1430959_at - -
          Mouse Expression Set 430 1430959_PM_at - -
          Mouse Expression Set 430 1453713_PM_s_at - -
          Mouse Expression Set 430 1453713_s_at - -
          Murine Genome U74v2 Set 133892_at - -
          Murine Genome U74v2 Set 133892_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430959_at - -
          Affymetrix HT Mouse Genome 430B Array 1453713_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1453713_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1430959_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717117 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline