Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01135354_m1
          See other 1700018A04RIK GE Assays ›
          Gene Symbol:
          1700018A04Rik
          Gene Name:
          RIKEN cDNA 1700018A04 gene
          Gene Aliases:
          4930551G20Rik, 4933439J11Rik
          Chromosome Location:
          Chr.13: 31565492 - 31582513 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01135354_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700018A04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029439.1 5 - 6 588 74
          GenBank mRNA AK006076.1 - 5 - 6 581 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 11377 Gm11377 OTTMUSG00000000685 Anti-sense Chr. 13 - 31564117 - 31567391 on NCBI Build GRCm38
          Downstream forkhead box F2 Foxf2 FREAC2,
          Fkh20,
          LUN
          58.4kb Anti-sense Chr. 13 - 31625816 - 31631406 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001747.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P111449 AK017122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P111449 AK142058.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154142 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151049 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133702 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P111449 AK017122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P111449 AK142058.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827644 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827613 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827672 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07827606 - -
          Whole Mouse Genome G4122A A_52_P115369 AK017122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P115369 AK142058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P115369 AK016091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P115369 BC049749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P115369 NR_029439.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111449 AK017122.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111449 AK142058.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133702 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10611051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11122330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10675663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10922231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10456768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10899349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10540080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10970950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11105147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11118542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10618065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10884410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10614417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10924510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10614430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10618063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10899347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10951924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10922229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10884402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10884401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11010405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28923915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408527 - -
          Affymetrix HT MG-430 PM Array Plate 1431567_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408525 - -
          Mouse Genome 430 2.0 Array 1431567_at - -
          Mouse Genome 430 2.0 Array 1431567_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300010858.mm.1 NR_029439.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010858.mm.1 NR_029439.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010858.mm.1 TC1300001747.mm.1 MTA TC
          Mouse Expression Set 430 1431567_at - -
          Mouse Expression Set 430 1431567_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431567_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431567_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105570095 - -
          Illumina mouse-6 v1.0 expression beadchip 105390368 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2545658 - -
          Illumina Sentrix mouse-6.v1 bead array 105390368 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105390368 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2545658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline