Search
Search
| Gene Symbol: Rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011302.3 | NP_035432.3 | 4 - 5 | 499 | 64 | |
| GenBank mRNA | AF073780.1 | - | 4 - 5 | 362 | 64 | |
| AF084561.1 | - | 4 - 5 | 370 | 64 | ||
| AJ011381.1 | - | 4 - 5 | 350 | 64 | ||
| BC046422.1 | - | 4 - 5 | 499 | 5401012 | 64 | |
Set Membership: |
|