Search
Search

Bcl9l| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030256.2 | NP_084532.2 | 3 - 4 | 629 | 64 | |
| GenBank mRNA | AB128033.1 | - | 2 - 3 | 415 | 64 | |
| AK155681.1 | - | 4 - 5 | 589 | 64 | ||
| AY296058.1 | - | 3 - 4 | 629 | 64 | ||
| BC082304.1 | - | 2 - 3 | 763 | 6856460 | 64 | |
Set Membership: |
|