Search
Search

Ddx21| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019553.2 | NP_062426.2 | 2 - 3 | 835 | 98 | |
| GenBank mRNA | AF159131.3 | - | 2 - 3 | 796 | 98 | |
| AK010281.1 | - | 2 - 3 | 715 | 98 | ||
| AK145578.1 | - | 2 - 3 | 819 | 98 | ||
| AK160095.1 | - | 2 - 3 | 809 | 98 | ||
| BC030895.1 | - | 1 - 2 | 267 | 98 | ||
| BC043655.1 | - | 2 - 3 | 807 | 5363948 | 98 | |
| BC059237.1 | - | 2 - 3 | 795 | 6412782 | 98 | |
| BC060220.1 | - | 2 - 3 | 787 | 98 | ||
Set Membership: |
|