Search
Search
| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113401.1 | NP_001106872.1 | 3 - 4 | 445 | 74 | |
| NM_134111.2 | NP_598872.1 | 3 - 4 | 435 | 74 | ||
| GenBank mRNA | AK076871.1 | - | 4 - 5 | 524 | 74 | |
| AY034479.1 | - | 3 - 4 | 452 | 74 | ||
| AY049021.1 | - | 3 - 4 | 391 | 74 | ||
| BC004721.1 | - | 3 - 4 | 434 | 3582359 | 74 | |
| BC056626.1 | - | 3 - 4 | 435 | 5008372 | 74 | |
Set Membership: |
|