Search
Search
| Gene Symbol: Nfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008688.3 | NP_032714.1 | 8 - 9 | 1341 | 100 | |
| GenBank mRNA | AK087539.1 | - | 8 - 9 | 1326 | 100 | |
| AK155258.1 | - | 8 - 9 | 1343 | 100 | ||
| Y07692.1 | - | 8 - 9 | 1300 | 100 | ||
| Y07693.1 | - | 8 - 9 | 1646 | 100 | ||
Set Membership: |
|