Search
Search

Adarb1| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024837.2 | NP_001020008.1 | 9 - 10 | 2175 | 60 | |
| NM_130895.3 | NP_570965.2 | 9 - 10 | 2145 | 60 | ||
| NR_004429.1 | 9 - 10 | 2222 | 60 | |||
| NR_021486.1 | 9 - 10 | 2192 | 60 | |||
| GenBank mRNA | AF291049.1 | - | 8 - 9 | 1980 | 60 | |
| AF403106.2 | - | 9 - 10 | 2097 | 60 | ||
| AF403107.2 | - | 9 - 10 | 2127 | 60 | ||
| AF403108.2 | - | 9 - 10 | 2144 | 60 | ||
| AF403109.2 | - | 9 - 10 | 2174 | 60 | ||
| AF525421.1 | - | 10 - 11 | 2170 | 60 | ||
| AK141777.1 | - | 9 - 10 | 2147 | 60 | ||
| AK147298.1 | - | 10 - 11 | 2254 | 60 | ||
| AY162454.1 | - | 10 - 11 | 2170 | 60 | ||
| BC040200.1 | - | 5 - 6 | 730 | 60 | ||
Set Membership: |
|