Search
Search

Pcsk4| Gene Symbol: Pcsk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008793.2 | NP_032819.1 | 12 - 13 | 1592 | 116 | |
| GenBank mRNA | AK161504.1 | - | 10 - 11 | 1851 | 116 | |
| D01093.1 | - | 12 - 13 | 1592 | 116 | ||
Set Membership: |
|