Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01150061_m1
          See other DOS GE Assays ›
          Gene Symbol:
          Dos
          Gene Name:
          downstream of Stk11
          Gene Aliases:
          9630008F14, R29144/1
          Chromosome Location:
          Chr.10: 80130434 - 80140136 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01150061_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195268.1 NP_001182197.1 8 - 9 1134 126
          NR_036582.1 10 - 11 1470 126
          GenBank mRNA AK035824.1 - 2 - 3 285 126
          BC052075.1 - 9 - 10 1188 126
          BC078442.1 - 4 - 5 689 126
          BC100552.1 - 3 - 4 517 5687195 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002491.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123101 BC100552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123101 BC052075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123101 BC078442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123101 BC026893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123101 BC039779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC100552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC052075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC078442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC026893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC039779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC100552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC052075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC078442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC026893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436817 BC039779.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370757 - -
          Affymetrix Mouse Gene 1.1 ST Array 10370754 - -
          Affymetrix Mouse Clariom S Array PSR1000018349.mm.1 NM_001195268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018349.mm.1 NM_001195268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018349.mm.1 NR_036582.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018349.mm.1 NR_036582.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018349.mm.1 TC1000002491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018351.mm.1 NM_001195268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018351.mm.1 NM_001195268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018351.mm.1 NR_036582.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018351.mm.1 NR_036582.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018351.mm.1 TC1000002491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018355.mm.1 NM_001195268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018355.mm.1 NM_001195268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018355.mm.1 NR_036582.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018355.mm.1 NR_036582.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018355.mm.1 TC1000002491.mm.1 MTA TC
          Mouse Expression Set 430 1438370_x_at - -
          Mouse Expression Set 430 1438370_PM_x_at - -
          Mouse Expression Set 430 1433494_at - -
          Mouse Expression Set 430 1433494_PM_at - -
          Murine Genome U74v2 Set 141128_at - -
          Murine Genome U74v2 Set 141128_PM_at - -
          Mouse Genome 430A 2.0 Array 1438370_x_at - -
          Mouse Genome 430A 2.0 Array 1433494_PM_at - -
          Mouse Genome 430A 2.0 Array 1433494_at - -
          Mouse Genome 430A 2.0 Array 1438370_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104560082 - -
          MouseWG-6 v2 Expression BeadChip 100670075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2668236 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline