Search
Search
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 17 - 18 | 1954 | 84 | |
| GenBank mRNA | AF237669.1 | - | 17 - 18 | 1913 | 84 | |
| AF239661.1 | - | 17 - 18 | 1955 | 84 | ||
| AK004876.1 | - | 14 - 15 | 1982 | 84 | ||
| AK131140.1 | - | 1460 | 84 | |||
| AK134654.1 | - | 1936 | 84 | |||
| AK165023.1 | - | 14 - 15 | 2343 | 84 | ||
| AK165746.1 | - | 15 - 16 | 2246 | 84 | ||
Set Membership: |
|