Search
Search

Ppcdc| Gene Symbol: Ppcdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176831.4 | NP_789801.1 | 3 - 4 | 293 | 68 | |
| GenBank mRNA | AK036042.1 | - | 3 - 4 | 276 | 68 | |
| AK038628.1 | - | 3 - 4 | 271 | 68 | ||
| BC004779.1 | - | 3 - 4 | 286 | 68 | ||
| BC052928.1 | - | 2 - 3 | 242 | 5255304 | 68 | |
| BC089351.1 | - | 2 - 3 | 257 | 30603115 | 68 | |
Set Membership: |
|