Search
Search
| Gene Symbol: Tex9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009359.4 | NP_033385.2 | 2 - 3 | 108 | 127 | |
| GenBank mRNA | AB000619.1 | - | 3 - 4 | 356 | 127 | |
| AK008505.1 | - | 2 - 3 | 106 | 127 | ||
| AK012189.1 | - | 2 - 3 | 107 | 127 | ||
| AK018568.1 | - | 106 | 127 | |||
| AK032490.1 | - | 1 - 2 | 215 | 127 | ||
| AK141174.1 | - | 1 - 2 | 218 | 127 | ||
| BC141033.1 | - | 2 - 3 | 250 | 9055645 | 127 | |
| BC145184.1 | - | 2 - 3 | 250 | 127 | ||
| BC145185.1 | - | 2 - 3 | 250 | 9053736 | 127 | |
| BC145186.1 | - | 2 - 3 | 250 | 9053737 | 127 | |
Set Membership: |
|