Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01152080_m1
          See other CCPG1 GE Assays ›
          Gene Symbol:
          Ccpg1
          Gene Name:
          cell cycle progression 1
          Gene Aliases:
          1700030B06Rik, 1810073J13Rik, 9430028F23Rik, AI426686, AI875170, CPR8, D9Ertd392e
          Chromosome Location:
          Chr.9: 72985427 - 73016340 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01152080_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccpg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114328.2 NP_001107800.1 1 - 2 171 59
          NM_001286544.1 NP_001273473.1 1 - 2 171 59
          NM_001286545.1 NP_001273474.1 1 - 2 171 59
          NM_001286546.1 NP_001273475.1 1 - 2 171 59
          NM_028181.5 NP_082457.3 1 - 2 171 59
          GenBank mRNA AK007973.1 - 1 - 2 68 59
          AK032645.1 - 1 - 2 57 59
          AK076325.2 - 1 - 2 98 59
          AK128894.1 - 1 - 2 97 59
          AK134675.1 - 1 - 2 68 59
          BC043049.1 - 1 - 2 70 5704185 59
          BC082602.1 - 1 - 2 72 30663407 59

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK007973.1, AK076325.2, BC043049.1, BC082602.1, NM_001114328.2, NM_001286544.1, NM_001286545.1, NM_001286546.1, NM_028181.5 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccpg1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          positive regulation of cell proliferation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of Rho guanyl-nucleotide exchange factor activity
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 NM_001114328.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 NM_001286544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 NM_001286545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 NM_001286546.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 NM_028181.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007613.mm.1 TC0900003326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 NM_001114328.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 NM_001286544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 NM_001286545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 NM_001286546.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 NM_028181.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007612.mm.1 TC0900003326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 NM_001114328.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 NM_001286544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 NM_001286545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 NM_001286546.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 NM_028181.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007619.mm.1 TC0900003326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 NM_001114328.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 NM_001286544.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 NM_001286545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 NM_001286546.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 NM_028181.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007622.mm.1 TC0900003326.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 741992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline