| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600003165.mm.1 |
NM_201370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003165.mm.1 |
NM_201370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003165.mm.1 |
TC0600000409.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003163.mm.1 |
NM_201370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003163.mm.1 |
NM_201370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003163.mm.1 |
TC0600000409.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003164.mm.1 |
NM_201370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003164.mm.1 |
NM_201370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003164.mm.1 |
TC0600000409.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600003162.mm.1 |
NM_201370.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003162.mm.1 |
NM_201370.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600003162.mm.1 |
TC0600000409.mm.1 |
MTA TC |