Search
Search

Cdkl5| Gene Symbol: Cdkl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024624.2 | NP_001019795.1 | 8 - 9 | 812 | 79 | |
| GenBank mRNA | AK052380.1 | - | 8 - 9 | 809 | 79 | |
| AK139220.1 | - | 8 - 9 | 784 | 79 | ||
Set Membership: |
|