Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01156900_m1
          See other WNT2 GE Assays ›
          Gene Symbol:
          Wnt2
          Gene Name:
          wingless-type MMTV integration site family, member 2
          Gene Aliases:
          2610510E18Rik, Int1l1, Irp, Mirp, Wnt-2, Wnt2a
          Chromosome Location:
          Chr.6: 17985474 - 18032751 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01156900_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023653.5 NP_076142.3 1 - 2 244 78
          GenBank mRNA AK012093.1 - 1 - 2 160 78
          AK045120.1 - 1 - 2 156 78
          AK167642.1 - 1 - 2 160 78
          BC026373.1 - 1 - 2 139 4162686 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001902.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wnt2

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          positive regulation of mesenchymal cell proliferation
          cell-cell signaling
          multicellular organism development
          positive regulation of cell proliferation
          Wnt signaling pathway
          neuron differentiation
          cell fate commitment
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          atrial cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          canonical Wnt signaling pathway
          cardiac epithelial to mesenchymal transition
          lung induction
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          labyrinthine layer blood vessel development
          positive regulation of canonical Wnt signaling pathway
          receptor binding
          frizzled binding
          cytokine activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600014897.mm.1 NM_023653.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014897.mm.1 NM_023653.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014897.mm.1 TC0600001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600014894.mm.1 NM_023653.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014894.mm.1 NM_023653.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014894.mm.1 TC0600001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600014895.mm.1 NM_023653.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014895.mm.1 NM_023653.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014895.mm.1 TC0600001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600014898.mm.1 NM_023653.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014898.mm.1 NM_023653.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014898.mm.1 TC0600001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600014896.mm.1 NM_023653.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014896.mm.1 NM_023653.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0600014896.mm.1 TC0600001902.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline