Search
Search

Rps6ka3| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 16 - 17 | 1783 | 97 | |
| GenBank mRNA | AK079102.1 | - | 1876 | 97 | ||
| AK154778.1 | - | 16 - 17 | 1695 | 97 | ||
| AY083469.1 | - | 16 - 17 | 1445 | 97 | ||
| BC150156.1 | - | 16 - 17 | 1448 | 97 | ||
| BC150478.1 | - | 16 - 17 | 1448 | 97 | ||
Set Membership: |
|