Search
Search

Rps6ka3| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148945.2 | NP_683747.1 | 4 - 5 | 670 | 144 | |
| GenBank mRNA | AK079102.1 | - | 763 | 144 | ||
| AK154778.1 | - | 4 - 5 | 583 | 144 | ||
| AY083469.1 | - | 4 - 5 | 332 | 144 | ||
Set Membership: |
|