Search
Search
| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291809.1 | NP_001278738.1 | 7 - 8 | 1120 | 141 | |
| NM_007882.3 | NP_031908.3 | 7 - 8 | 1120 | 141 | ||
| GenBank mRNA | BC144845.1 | - | 7 - 8 | 1105 | 141 | |
| Y11169.1 | - | 7 - 8 | 1120 | 141 | ||
Set Membership: |
|