Search
Search
| Gene Symbol: Nrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167891.1 | NP_001161363.1 | 2 - 3 | 910 | 81 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BI412864.1 | - | Chr. - - [-] on Build | 81 | |||
Set Membership: |
|