Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01158117_m1
          See other TMEM173 GE Assays ›
          Gene Symbol:
          Tmem173
          Gene Name:
          transmembrane protein 173
          Gene Aliases:
          2610307O08Rik, ERIS, MPYS, Mita, STING
          Chromosome Location:
          Chr.18: 35733678 - 35740554 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01158117_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem173
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289591.1 NP_001276520.1 4 - 5 763 104
          NM_001289592.1 NP_001276521.1 5 - 6 730 104
          NM_028261.1 NP_082537.1 5 - 6 853 104
          GenBank mRNA AK012006.1 - 5 - 6 800 104
          AK077788.1 - 5 - 6 754 104
          AK089405.1 - 5 - 6 831 104
          AK146284.1 - 5 - 6 726 104
          AK153868.1 - 5 - 6 724 104
          AK157370.1 - 4 - 5 665 104
          AK158458.1 - 5 - 6 639 104
          AK170724.1 - 5 - 6 831 104
          AK171065.1 - 5 - 6 817 104
          AK171203.1 - 5 - 6 817 104
          AK171612.1 - 5 - 6 784 104
          AK172314.1 - 4 - 5 595 104
          AX775911.1 - 5 - 6 740 104
          BC027757.1 - 3 - 4 497 104
          BC046640.1 - 4 - 5 688 5067343 104
          DQ910493.1 - 5 - 6 651 104
          FJ222242.1 - 3 - 4 515 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001181.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of innate immune response
          positive regulation of defense response to virus by host
          immune system process
          apoptotic process
          positive regulation of protein binding
          positive regulation of type I interferon production
          interferon-beta production
          positive regulation of protein import into nucleus, translocation
          cellular response to interferon-beta
          positive regulation of transcription factor import into nucleus
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          cellular response to exogenous dsRNA
          nucleotide binding
          protein binding
          transcription factor binding
          protein kinase binding
          ubiquitin protein ligase binding
          cyclic-di-GMP binding
          identical protein binding
          protein homodimerization activity
          cyclic-GMP-AMP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC027757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC046640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC027757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 BC046640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK171612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13326 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159411 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC027757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC046640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK172314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC027757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 BC046640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK171612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03449407 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159411 - -
          Whole Mouse Genome G4122A A_51_P240801 BC027757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC046640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK172314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC027757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 BC046640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK171612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240801 AK172314.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13688687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13640983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14107177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31619861 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458321 - -
          Affymetrix HT MG-430 PM Array Plate 1427911_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447621_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458314 - -
          Mouse Genome 430 2.0 Array 1427911_at - -
          Mouse Genome 430 2.0 Array 1447621_PM_s_at - -
          Mouse Genome 430 2.0 Array 1447621_s_at - -
          Mouse Genome 430 2.0 Array 1427911_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72512_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72512_at - -
          Affymetrix Mouse Clariom S Array PSR1800008718.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008718.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008718.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008718.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008718.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008721.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008715.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008715.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008715.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008715.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008715.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008714.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008723.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008722.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008719.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008719.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008719.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008719.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008716.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008716.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008716.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008716.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008716.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008720.mm.1 TC1800001181.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008717.mm.1 NM_001289591.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008717.mm.1 NM_001289592.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008717.mm.1 NM_028261.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008717.mm.1 NM_028261.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008717.mm.1 TC1800001181.mm.1 MTA TC
          Mouse Expression Set 430 1427911_at - -
          Mouse Expression Set 430 1427911_PM_at - -
          Mouse Expression Set 430 1447621_PM_s_at - -
          Mouse Expression Set 430 1447621_s_at - -
          Murine Genome U74v2 Set 168500_s_at - -
          Murine Genome U74v2 Set 168500_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427911_PM_at - -
          Mouse Genome 430A 2.0 Array 1427911_at - -
          Affymetrix HT Mouse Genome 430A Array 1427911_at - -
          Affymetrix HT Mouse Genome 430A Array 1427911_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447621_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447621_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 503137 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736197 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736197 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline