Search
Search

Pot1a| Gene Symbol: Pot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133931.4 | NP_598692.1 | 7 - 8 | 925 | 68 | |
| GenBank mRNA | AK036052.1 | - | 7 - 8 | 926 | 68 | |
| AK040864.1 | - | 7 - 8 | 918 | 68 | ||
| AK085041.1 | - | 7 - 8 | 916 | 68 | ||
| AK135346.1 | - | 7 - 8 | 895 | 68 | ||
| AK137376.1 | - | 7 - 8 | 895 | 68 | ||
| BC016121.1 | - | 7 - 8 | 887 | 4501733 | 68 | |
Set Membership: |
|