Search
Search
| Gene Symbol: Ace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130513.1 | NP_001123985.1 | 9 - 10 | 1393 | 80 | |
| NM_027286.4 | NP_081562.2 | 8 - 9 | 1245 | 80 | ||
| GenBank mRNA | AB053181.1 | - | 8 - 9 | 1249 | 80 | |
| AK135357.1 | - | 3 - 4 | 427 | 80 | ||
| AK141604.1 | - | 8 - 9 | 1094 | 80 | ||
| BC026801.1 | - | 8 - 9 | 1238 | 4236529 | 80 | |
| EF408739.1 | - | 9 - 10 | 1354 | 80 | ||
| EF408740.1 | - | 9 - 10 | 1354 | 80 | ||
| GQ262785.1 | - | 8 - 9 | 1063 | 80 | ||
Set Membership: |
|