Search
Search
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019776.2 | NP_062750.2 | 1 - 2 | 299 | 133 | |
| GenBank mRNA | AB021491.1 | - | 1 - 2 | 256 | 133 | |
| AK034914.1 | - | 1 - 2 | 247 | 133 | ||
| AK088028.1 | - | 1 - 2 | 258 | 133 | ||
| AK133837.1 | - | 1 - 2 | 305 | 133 | ||
| AK159837.1 | - | 1 - 2 | 264 | 133 | ||
| AK161591.1 | - | 1 - 2 | 253 | 133 | ||
| AK162432.1 | - | 1 - 2 | 240 | 133 | ||
| AK167576.1 | - | 1 - 2 | 289 | 133 | ||
| BB613775.1 | - | 1 - 2 | 299 | 133 | ||
| BC007126.1 | - | 1 - 2 | 246 | 3592548 | 133 | |
Set Membership: |
|