Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01159156_g1
          See other HSD3B7 GE Assays ›
          Gene Symbol:
          Hsd3b7
          Gene Name:
          hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
          Gene Aliases:
          AI195443, BB098564
          Chromosome Location:
          Chr.7: 127800609 - 127803802 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01159156_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsd3b7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040684.1 NP_001035774.1 6 - 7 750 68
          NM_133943.2 NP_598704.2 6 - 7 802 68
          GenBank mRNA AF277718.1 - 6 - 7 738 68
          AK077476.1 - 5 - 6 853 68
          AK150786.1 - 1 - 2 174 68
          BC012954.1 - 5 - 6 603 68
          BC132605.1 - 6 - 7 736 40130882 68
          BC138589.1 - 6 - 7 736 8861609 68
          CB951288.1 - 3 - 4 359 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004630.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsd3b7

          Panther Classification:

          Molecular Function -

          dehydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          steroid biosynthetic process
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          3-beta-hydroxy-delta5-steroid dehydrogenase activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase activity
          3-ketoglucose-reductase activity
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982762 BC012954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982762 BC018525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982762 AK077476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982762 AK150786.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982762 BC006822.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683408 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982762 BC012954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982762 BC018525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982762 AK077476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982762 AK150786.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982762 BC006822.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435911 BC138589.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435911 BC012954.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435911 BC018525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435911 BC132605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P435911 AK077476.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25507396 - -
          Whole Mouse Genome G4122A A_51_P435911 BC138589.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435911 BC012954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435911 BC018525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435911 BC132605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P435911 AK077476.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06524192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06059542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06524206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25507442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06524192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25507417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06059526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557789 - -
          Affymetrix HT MG-430 PM Array Plate 1416968_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568296 - -
          Affymetrix Mouse Gene 1.1 ST Array 10557782 - -
          Mouse Genome 430 2.0 Array 1416968_a_at - -
          Mouse Genome 430 2.0 Array 1416968_PM_a_at - -
          Mouse Genome 430 2.0 Array 1442912_at - -
          Mouse Genome 430 2.0 Array 1442912_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101502_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101502_at - -
          Affymetrix Mouse Clariom S Array PSR0700016077.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016077.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016077.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016077.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016077.mm.1 NM_133943.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016079.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016079.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016079.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016079.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016079.mm.1 NM_133943.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016081.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016081.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016081.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016081.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016081.mm.1 NM_133943.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016075.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016075.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016075.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016075.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016075.mm.1 NM_133943.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016082.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016082.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016082.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016082.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016082.mm.1 NM_133943.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016076.mm.1 TC0700004630.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016076.mm.1 NM_001040684.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016076.mm.1 NM_001040684.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016076.mm.1 NM_133943.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016076.mm.1 NM_133943.2 Refseq
          Mouse Expression Set 430 1416968_PM_a_at - -
          Mouse Expression Set 430 1416968_a_at - -
          Murine Genome U74v2 Set 160104_PM_at - -
          Murine Genome U74v2 Set 160104_at - -
          Mouse Genome 430A 2.0 Array 1416968_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416968_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416968_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416968_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410129 - -
          Illumina mouse-6 v1.0 expression beadchip 1410129 - -
          Illumina mouse-6 v1.0 expression beadchip 103120619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260456 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260456 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227060 - -
          mouse-6 v1.1 (Illumina) scl00101502.1_299-S - -
          mouse-6 v1.1 (Illumina) scl30633.2.1_192-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410129 - -
          Illumina Sentrix mouse-6.v1 bead array 103120619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00101502.1_299-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410129 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103120619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1260456 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3114124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260456 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3114124 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00101502.1_299-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline