Search
Search

Atp6v1g2| Gene Symbol: Atp6v1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023179.3 | NP_075668.1 | 1 - 2 | 255 | 85 | |
| GenBank mRNA | AB076405.1 | - | 1 - 2 | 310 | 85 | |
| AK005109.1 | - | 1 - 2 | 136 | 85 | ||
| AK011945.1 | - | 1 - 2 | 98 | 85 | ||
| BC020190.1 | - | 1 - 2 | 251 | 4505711 | 85 | |
| BC062380.1 | - | 1 - 2 | 243 | 6468413 | 85 | |
| BU558667.1 | - | 1 - 2 | 258 | 85 | ||
Set Membership: |
|