Search
Search

Spon1| Gene Symbol: Spon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145584.2 | NP_663559.1 | 10 - 11 | 1639 | 63 | |
| GenBank mRNA | AK146506.1 | - | 10 - 11 | 1463 | 63 | |
| AK173021.1 | - | 10 - 11 | 1306 | 63 | ||
| BC020531.1 | - | 10 - 11 | 1509 | 4221758 | 63 | |
| BC030339.1 | - | 1 - 2 | 93 | 63 | ||
Set Membership: |
|