Search
Search

Stat6| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009284.2 | NP_033310.2 | 6 - 7 | 818 | 68 | |
| GenBank mRNA | AF481808.1 | - | 5 - 6 | 533 | 68 | |
| AF481809.1 | - | 5 - 6 | 533 | 68 | ||
| AK149563.1 | - | 6 - 7 | 778 | 68 | ||
| AK163053.1 | - | 6 - 7 | 819 | 68 | ||
| BC127054.1 | - | 6 - 7 | 608 | 40109712 | 68 | |
Set Membership: |
|