Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01160713_m1
          See other TCAF3 GE Assays ›
          Gene Symbol:
          Tcaf3
          Gene Name:
          TRPM8 channel-associated factor 3
          Gene Aliases:
          Eapa2, Fam115e
          Chromosome Location:
          Chr.6: 42587213 - 42597372 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01160713_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcaf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203396.1 NP_981933.1 6 - 7 2570 65
          GenBank mRNA AY528666.1 - 6 - 7 2570 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002176.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcaf3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959041 NM_203396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959041 AY528666.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130603 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121154 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172110 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959041 NM_203396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959041 AY528666.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24399209 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121154 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138905 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172110 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121154 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507766 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507779 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507780 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05507764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24399166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05213613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05092350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05507776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05036831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05213629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05213610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05648188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05179034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05487728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05213617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05329453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24399210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544448 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544444 - -
          Affymetrix Mouse Clariom S Array PSR0600017140.mm.1 NM_203396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017140.mm.1 NM_203396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017140.mm.1 TC0600002176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017139.mm.1 NM_203396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017139.mm.1 NM_203396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017139.mm.1 TC0600002176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017141.mm.1 NM_203396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017141.mm.1 NM_203396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017141.mm.1 TC0600002176.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 648448 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5130500 - -
          Illumina mouse-6 v1.0 expression beadchip 5130500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692367 - -
          mouse-6 v1.1 (Illumina) scl0403088.1_170-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0403088.1_170-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692367 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline