Search
Search

Sema5a| Gene Symbol: Sema5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009154.2 | NP_033180.2 | 4 - 5 | 879 | 85 | |
| GenBank mRNA | AK031231.1 | - | 4 - 5 | 872 | 85 | |
| AK039101.1 | - | 4 - 5 | 865 | 85 | ||
| AK043386.1 | - | 4 - 5 | 591 | 85 | ||
| AK045236.1 | - | 4 - 5 | 883 | 85 | ||
| AK046502.1 | - | 4 - 5 | 655 | 85 | ||
| AK141612.1 | - | 4 - 5 | 823 | 85 | ||
| AK143026.1 | - | 4 - 5 | 890 | 85 | ||
| AK157706.1 | - | 4 - 5 | 862 | 85 | ||
| AK165206.1 | - | 4 - 5 | 1180 | 85 | ||
| BC065137.1 | - | 3 - 4 | 348 | 30548165 | 85 | |
| X97817.1 | - | 2 - 3 | 248 | 85 | ||
Set Membership: |
|