Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01163016_s1
          See other OLFR699 GE Assays ›
          Gene Symbol:
          Olfr699
          Gene Name:
          olfactory receptor 699
          Gene Aliases:
          MOR283-10P
          Chromosome Location:
          Chr.7: 106790049 - 106790999 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01163016_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr699
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011862.1 NP_001011862.1 1 452 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr699

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032014 NM_001011862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032014 NM_001011862.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396598 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06301693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06585934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06585929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05955737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06585943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06585930 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06301670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06301680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06301675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06301673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06585941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396595 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566600 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566599 - -
          Affymetrix Mouse Clariom S Array PSR0700033447.mm.1 NM_001011862.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033447.mm.1 NM_001011862.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033447.mm.1 TC0700004018.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616018 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2789964 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2789964 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline