Search
Search
| Gene Symbol: Cln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146311.1 | NP_001139783.1 | 11 - 12 | 1092 | 91 | |
| NM_009907.3 | NP_034037.3 | 9 - 10 | 980 | 91 | ||
| GenBank mRNA | AK078976.1 | - | 9 - 10 | 964 | 91 | |
| AK134036.1 | - | 12 - 13 | 1615 | 91 | ||
| BC058753.1 | - | 10 - 11 | 916 | 6477909 | 91 | |
| BC060306.1 | - | 8 - 9 | 862 | 5250894 | 91 | |
| BC080759.1 | - | 10 - 11 | 874 | 5353776 | 91 | |
| U47106.1 | - | 9 - 10 | 930 | 91 | ||
| U68064.1 | - | 9 - 10 | 950 | 91 | ||
| U92811.1 | - | 9 - 10 | 899 | 91 | ||
Set Membership: |
|