Search
Search
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009464.3 | NP_033490.1 | 3 - 4 | 532 | 74 | |
| GenBank mRNA | AB008216.1 | - | 2 - 3 | 335 | 74 | |
| AB010742.1 | - | 3 - 4 | 532 | 74 | ||
| AB013132.1 | - | 1 - 2 | 88 | 74 | ||
| AF030164.1 | - | 2 - 3 | 373 | 74 | ||
| AF053352.1 | - | 3 - 4 | 504 | 74 | ||
| BC139430.1 | - | 2 - 3 | 400 | 8862452 | 74 | |
| BC139431.1 | - | 2 - 3 | 400 | 8862453 | 74 | |
Set Membership: |
|