Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01163611_m1
          See other CTHRC1 GE Assays ›
          Gene Symbol:
          Cthrc1
          Gene Name:
          collagen triple helix repeat containing 1
          Gene Aliases:
          1110014B07Rik
          Chromosome Location:
          Chr.15: 39076851 - 39087121 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01163611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cthrc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026778.2 NP_081054.1 2 - 3 449 75
          GenBank mRNA AK003674.1 - 2 - 3 445 75
          AK076498.1 - 2 - 3 452 75
          AK151282.1 - 2 - 3 450 75
          BC131907.1 - 2 - 3 425 40130184 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000300.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell migration
          positive regulation of protein binding
          positive regulation of osteoblast proliferation
          ossification involved in bone remodeling
          positive regulation of osteoblast differentiation
          Wnt signaling pathway, planar cell polarity pathway
          inner ear receptor stereocilium organization
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          establishment of planar polarity involved in neural tube closure
          frizzled binding
          protein binding
          Wnt-protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151912 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK151282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 NM_026778.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK003674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK076498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK151282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 NM_026778.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK003674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P453785 AK076498.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 AK080681.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 AK168592.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 BC016216.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 BC025115.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 BC078460.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 NM_001110254.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858145 NM_177358.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158432 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150796 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK151282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 NM_026778.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK003674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK076498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK151282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 NM_026778.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK003674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P453785 AK076498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211702 AK090258.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158432 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150796 - -
          Whole Mouse Genome G4122A A_52_P453785 AK151282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 NM_026778.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 AK003674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 AK076498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 AK151282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 NM_026778.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 AK003674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453785 AK076498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211702 AK090258.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158432 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151915 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151913 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30151912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30152018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30152014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30152016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30152015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30152021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30151959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12315650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12238702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12007162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12465891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12315651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30151911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12076900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311678 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10423837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423841 - -
          Affymetrix HT MG-430 PM Array Plate 1452968_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10423836 - -
          Mouse Genome 430 2.0 Array 1452968_at - -
          Mouse Genome 430 2.0 Array 1452968_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68588_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68588_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500002155.mm.1 NM_026778.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002155.mm.1 NM_026778.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002155.mm.1 TC1500000300.mm.1 MTA TC
          Mouse Expression Set 430 1452968_PM_at - -
          Mouse Expression Set 430 1452968_at - -
          Affymetrix HT Mouse Genome 430B Array 1452968_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452968_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 864310 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2713401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2713401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225073 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2713401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline