Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171003.1 | NP_001164474.1 | 34 - 35 | 4206 | 77 | |
GenBank mRNA | AK031332.1 | - | 18 - 19 | 2210 | 77 | |
EU937529.1 | - | 34 - 35 | 4206 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |