Search
Search
| Gene Symbol: Irf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252601.1 | NP_001239530.1 | 5 - 6 | 899 | 63 | |
| GenBank mRNA | AK002830.1 | - | 5 - 6 | 897 | 63 | |
| AK158378.1 | - | 5 - 6 | 898 | 63 | ||
| AK158403.1 | - | 5 - 6 | 898 | 63 | ||
| DT923384.1 | - | 3 - 4 | 376 | 63 | ||
Set Membership: |
|