Search
Search

Irak3| Gene Symbol: Irak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028679.3 | NP_082955.2 | 6 - 7 | 771 | 92 | |
| GenBank mRNA | AF461763.1 | - | 6 - 7 | 751 | 92 | |
| AJ440757.2 | - | 6 - 7 | 693 | 92 | ||
| AK029057.1 | - | 6 - 7 | 772 | 92 | ||
| AK049210.1 | - | 6 - 7 | 802 | 92 | ||
| BC120829.1 | - | 6 - 7 | 732 | 40129752 | 92 | |
| BC120831.1 | - | 6 - 7 | 732 | 40129754 | 92 | |
| BC144835.1 | - | 6 - 7 | 732 | 92 | ||
Set Membership: |
|