Search
Search

Dhcr7| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007856.2 | NP_031882.1 | 6 - 7 | 1120 | 136 | |
| GenBank mRNA | AF057368.1 | - | 5 - 6 | 652 | 136 | |
| BC006854.1 | - | 4 - 5 | 634 | 3157987 | 136 | |
| CT010277.1 | - | 4 - 5 | 613 | 136 | ||
Set Membership: |
|