Search
Search

1700006J14Rik| Gene Symbol: 1700006J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011105.1 | - | 3 - 4 | 335 | 55 | |
| BC098228.1 | - | 3 - 4 | 341 | 55 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
LLP homolog, long-term synaptic facilitation (Aplysia) |
Llph | 1190005P17Rik | -139.2kb | Anti-sense | Chr. 10 - 120227060 - 120232071 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9230105E05 gene | 9230105E05Rik | 7.3kb | Anti-sense | Chr. 10 - 120389523 - 120392793 on NCBI Build GRCm38 |
Set Membership: |
|