Search
Search

Cdh12| Gene Symbol: Cdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008420.2 | NP_001008420.1 | 7 - 8 | 1450 | 72 | |
| GenBank mRNA | BC086655.1 | - | 7 - 8 | 1450 | 30542783 | 72 |
Set Membership: |
|