Search
Search

Rabep2| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030566.2 | NP_085043.2 | 9 - 10 | 1747 | 84 | |
| GenBank mRNA | AK089938.1 | - | 9 - 10 | 1452 | 84 | |
| AK144836.1 | - | 9 - 10 | 1433 | 84 | ||
| AK157832.1 | - | 9 - 10 | 1408 | 84 | ||
| BC004088.1 | - | 8 - 9 | 1493 | 3593722 | 84 | |
| BC015287.1 | - | 9 - 10 | 1755 | 4240442 | 84 | |
Set Membership: |
|