Search
Search

Arhgef5| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 6 - 7 | 3756 | 88 | |
| GenBank mRNA | AK037202.1 | - | 1 - 2 | 173 | 88 | |
| BC006829.1 | - | 1 - 2 | 147 | 88 | ||
| BC025127.1 | - | 1 - 2 | 43 | 88 | ||
Set Membership: |
|