Search
Search
| Gene Symbol: Gli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008130.2 | NP_032156.2 | 11 - 12 | 2085 | 110 | |
| GenBank mRNA | AK144851.1 | - | 11 - 12 | 1869 | 110 | |
| BC141135.1 | - | 11 - 12 | 1786 | 9055747 | 110 | |
| BC145445.1 | - | 11 - 12 | 1786 | 9054003 | 110 | |
| X95255.1 | - | 11 - 12 | 1825 | 110 | ||
Set Membership: |
|