Search
Search

Mkln1| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 9 - 10 | 993 | 100 | |
| GenBank mRNA | AK077541.1 | - | 9 - 10 | 988 | 100 | |
| BC013703.1 | - | 9 - 10 | 977 | 3668925 | 100 | |
| U72194.1 | - | 9 - 10 | 976 | 100 | ||
Set Membership: |
|