Search
Search

Pkhd1l1| Gene Symbol: Pkhd1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138674.2 | NP_619615.2 | 10 - 11 | 810 | 146 | |
| GenBank mRNA | AB055648.1 | - | 10 - 11 | 810 | 146 | |
| AY219182.1 | - | 10 - 11 | 810 | 146 | ||
Set Membership: |
|